Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FDFT1 SIRNA Assays ›
Gene Symbol:
FDFT1
Gene Name:
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases:
DGPT, ERG9, SQS, SS
Chromosome Location:
Chr.8: 11795573 - 11839309 on Build GRCh38
UniGene ID:
Hs.593928
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122387
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FDFT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287742.1
NP_001274671.1
9
1705
NM_001287743.1
NP_001274672.1
8
1507
NM_001287744.1
NP_001274673.1
7
1334
NM_001287745.1
NP_001274674.1
8
1413
NM_001287747.1
NP_001274676.1
7
1370
NM_001287748.1
NP_001274677.1
7
1032
NM_001287749.1
NP_001274678.1
7
1270
NM_001287750.1
NP_001274679.1
6
1272
NM_001287751.1
NP_001274680.1
6
1144
NM_001287756.1
NP_001274685.1
4
729
NM_004462.4
NP_004453.3
7
1216
XM_017013217.1
XP_016868706.1
9
1997
GenBank mRNA
AK057726.1
-
7
1334
AK098682.1
-
794
AK293545.1
-
7
1270
AK296043.1
-
6
959
AK297868.1
-
6
1215
AK300059.1
-
6
1034
AK300245.1
-
7
827
AK301617.1
-
4
590
AK315993.1
-
8
1283
AK316033.1
-
7
1237
AK316182.1
-
6
1232
AK316351.1
-
7
997
AK316531.1
-
7
982
AK316534.1
-
6
1144
BC003573.1
-
7
1063
3538137
BC009251.2
-
7
1063
2961623
BC029641.1
-
7
1026
4514761
BC034440.1
-
7
1026
BT006704.1
-
7
994
CR457033.1
-
7
994
L06070.1
-
7
1003
L06105.1
-
7
1038
S76822.1
-
7
1059
X69141.1
-
7
1085
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
Gene Ontology Categories:
Function(s)
Process(es)
steroid biosynthetic process
cholesterol biosynthetic process
isoprenoid biosynthetic process
farnesyl diphosphate metabolic process
oxidation-reduction process
farnesyl-diphosphate farnesyltransferase activity
oxidoreductase activity
squalene synthase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0