Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000038.5 | NP_000029.2 | 4 | 386 | ||
NM_001127510.2 | NP_001120982.1 | 5 | 494 | |||
NM_001127511.2 | NP_001120983.2 | 3 | 544 | |||
GenBank mRNA | AB210001.1 | - | 5 | 458 | ||
AK289567.1 | - | 4 | 448 | |||
AK294544.1 | - | 3 | 528 | |||
BC056268.1 | - | 4 | 357 | |||
BP230350.1 | - | 3 | 543 | |||
DA795663.1 | - | 4 | 386 | |||
M73548.1 | - | 3 | 339 | |||
M74088.1 | - | 3 | 319 |