Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000216.3 | NP_000207.2 | 6 | 897 | ||
XM_005274501.4 | XP_005274558.1 | 6 | 842 | |||
GenBank mRNA | BC137426.1 | - | 6 | 861 | 9021424 | |
BC137427.1 | - | 6 | 861 | 9021425 | ||
M97252.1 | - | 6 | 896 | |||
X60299.1 | - | 6 | 809 |