Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000216.3 | NP_000207.2 | 10, 11 | 1604 | ||
GenBank mRNA | BC137426.1 | - | 10, 11 | 1568 | 9021424 | |
BC137427.1 | - | 10, 11 | 1568 | 9021425 | ||
CD642906.1 | - | 3, 4 | 262 | |||
M97252.1 | - | 10, 11 | 1603 | |||
X60299.1 | - | 10, 11 | 1516 |