Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004475.2 | NP_004466.2 | 4 | 398 | ||
XM_005257950.3 | XP_005258007.1 | 4 | 429 | |||
XM_005257953.2 | XP_005258010.1 | 3 | 333 | |||
XM_017024394.1 | XP_016879883.1 | 4 | 326 | |||
XM_017024395.1 | XP_016879884.1 | 5 | 495 | |||
XM_017024396.1 | XP_016879885.1 | 4 | 332 | |||
GenBank mRNA | BC003683.1 | - | 4 | 310 | 3461759 | |
BC017292.1 | - | 3 | 220 | |||
BT019477.1 | - | 2 | 128 | |||
BT019478.1 | - | 2 | 128 | |||
CA390100.1 | - | 4 | 398 | |||
CR541784.1 | - | 2 | 128 | |||
CR542265.1 | - | 2 | 128 | |||
M60922.1 | - | 4 | 254 |