Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004475.2 | NP_004466.2 | 9 | 1163 | ||
XM_005257950.3 | XP_005258007.1 | 9 | 1194 | |||
XM_005257953.2 | XP_005258010.1 | 8 | 1098 | |||
XM_017024394.1 | XP_016879883.1 | 9 | 1091 | |||
XM_017024395.1 | XP_016879884.1 | 10 | 1260 | |||
XM_017024396.1 | XP_016879885.1 | 9 | 1097 | |||
GenBank mRNA | AK225491.1 | - | 2 | 193 | ||
BC003683.1 | - | 9 | 1075 | 3461759 | ||
BC017292.1 | - | 8 | 985 | |||
BT019477.1 | - | 7 | 893 | |||
BT019478.1 | - | 7 | 893 | |||
CR541784.1 | - | 7 | 893 | |||
CR542265.1 | - | 7 | 893 | |||
M60922.1 | - | 9 | 1019 |