Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004646.3 | NP_004637.1 | 4, 5 | 679 | ||
GenBank mRNA | AF035835.1 | - | 4, 5 | 523 | ||
AF190637.1 | - | 4, 5 | 523 | |||
EU642886.1 | - | 4, 5 | 523 | |||
EU642887.1 | - | 4, 5 | 523 |