Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143995.2 | NP_001137467.1 | 2 | 312 | ||
NM_001307951.1 | NP_001294880.1 | 2 | 345 | |||
NM_004811.2 | NP_004802.1 | 2 | 249 | |||
XM_006718750.1 | XP_006718813.1 | 2 | 345 | |||
XM_011545388.2 | XP_011543690.1 | 2 | 1016 | |||
XM_011545390.2 | XP_011543692.1 | 2 | 1326 | |||
XM_011545391.2 | XP_011543693.1 | 2 | 292 | |||
XM_011545393.2 | XP_011543695.1 | 2 | 292 | |||
XM_011545394.2 | XP_011543696.1 | 2 | 215 | |||
XM_017018579.1 | XP_016874068.1 | 2 | 292 | |||
GenBank mRNA | AF062075.1 | - | 2 | 197 | ||
AK223165.1 | - | 2 | 188 | |||
AK299264.1 | - | 2 | 326 | |||
AK300955.1 | - | 2 | 239 | |||
AK301093.1 | - | 2 | 188 | |||
AK313306.1 | - | 2 | 182 | |||
BC019035.2 | - | 2 | 171 | 4575069 | ||
BP261314.1 | - | 2 | 249 | |||
CR536531.1 | - | 1 | 104 | |||
DA363664.1 | - | 2 | 346 |