Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143995.2 | NP_001137467.1 | 3 | 393 | ||
NM_004811.2 | NP_004802.1 | 3 | 330 | |||
XM_006718750.1 | XP_006718813.1 | 3 | 426 | |||
XM_011545393.2 | XP_011543695.1 | 3 | 373 | |||
XM_011545394.2 | XP_011543696.1 | 3 | 296 | |||
XM_017018579.1 | XP_016874068.1 | 3 | 373 | |||
GenBank mRNA | AF062075.1 | - | 3 | 278 | ||
AK223165.1 | - | 3 | 269 | |||
AK300955.1 | - | 3 | 320 | |||
AK301093.1 | - | 3 | 269 | |||
AK313306.1 | - | 3 | 263 | |||
BC019035.2 | - | 3 | 252 | 4575069 | ||
BP261314.1 | - | 3 | 330 | |||
CR536531.1 | - | 2 | 185 | |||
DA363664.1 | - | 3 | 427 |