Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002817.3 | NP_002808.3 | 2 | 370 | ||
XM_011520235.2 | XP_011518537.1 | 2 | 503 | |||
GenBank mRNA | AB009398.1 | - | 2 | 197 | ||
AF083245.1 | - | 2 | 182 | |||
AF086708.1 | - | 2 | 128 | |||
AF107837.1 | - | 2 | 161 | |||
AK222895.1 | - | 2 | 201 | |||
AK295944.1 | - | 2 | 202 | |||
AK314733.1 | - | 2 | 632 | |||
BC001100.2 | - | 2 | 183 | 3506530 | ||
BC001747.1 | - | 2 | 151 | 3535807 | ||
BM456586.1 | - | 2 | 201 | |||
BT007307.1 | - | 2 | 128 | |||
BX647314.1 | - | 2 | 140 |