Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128930.2 | NP_001122402.1 | 9 | 1201 | ||
NM_004901.4 | NP_004892.1 | 9 | 1225 | |||
GenBank mRNA | AB002390.1 | - | 7 | 755 | ||
AF016032.1 | - | 9 | 1130 | |||
AJ131358.1 | - | 9 | 1122 | |||
AK300464.1 | - | 9 | 1133 | |||
BC034477.2 | - | 9 | 1165 | 4824559 |