Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242948.1 | NP_001229877.1 | 2 | 205 | ||
NM_017549.4 | NP_060019.2 | 2 | 720 | |||
GenBank mRNA | AJ250475.2 | - | 2 | 193 | ||
AK290885.1 | - | 2 | 414 | |||
AK309180.1 | - | 2 | 192 | |||
AY027862.2 | - | 2 | 720 | |||
BC000686.2 | - | 2 | 464 | |||
BC018299.1 | - | 2 | 612 | 3461888 | ||
DA938402.1 | - | 3 | 233 | |||
DB167672.1 | - | 2 | 206 |