Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242946.1 | NP_001229875.2 | 2 | 1203 | ||
NM_001242948.1 | NP_001229877.1 | 3 | 897 | |||
NM_017549.4 | NP_060019.2 | 3 | 1412 | |||
GenBank mRNA | AJ250475.2 | - | 3 | 885 | ||
AK290885.1 | - | 3 | 1106 | |||
AK309180.1 | - | 3 | 884 | |||
AY027862.2 | - | 3 | 1412 | |||
BC000686.2 | - | 3 | 1156 | |||
BC018299.1 | - | 3 | 1304 | 3461888 |