Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002819.4 | NP_002810.1 | 14 | 1605 | ||
NM_031990.3 | NP_114367.1 | 14 | 1584 | |||
NM_031991.3 | NP_114368.1 | 13 | 1527 | |||
XM_005259597.2 | XP_005259654.1 | 14 | 2108 | |||
XM_005259598.2 | XP_005259655.1 | 13 | 2030 | |||
GenBank mRNA | AB208910.1 | - | 11 | 4005 | ||
BC002397.2 | - | 14 | 1572 | 2821531 | ||
BC004383.1 | - | 13 | 1437 | 3615251 | ||
BC013694.1 | - | 14 | 1518 | 3863892 | ||
BC023219.1 | - | 14 | 1571 | |||
BT006819.1 | - | 13 | 1499 | |||
X60648.1 | - | 13 | 1449 | |||
X62006.1 | - | 13 | 1466 | |||
X65371.1 | - | 14 | 1523 | |||
X65372.1 | - | 14 | 1544 | |||
X66975.1 | - | 14 | 1586 |