Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318249.1 | NP_001305178.1 | 3 | 539 | ||
NM_001318250.1 | NP_001305179.1 | 4 | 631 | |||
NM_014298.4 | NP_055113.2 | 4 | 1075 | |||
NR_134536.1 | 3 | 568 | ||||
XM_005255223.3 | XP_005255280.2 | 5 | 1161 | |||
XM_017023101.1 | XP_016878590.1 | 5 | 1158 | |||
GenBank mRNA | AK090801.1 | - | 1 | 1610 | ||
AK293195.1 | - | 3 | 539 | |||
BC005060.1 | - | 4 | 912 | 2960170 | ||
BC010033.2 | - | 4 | 904 | 3506460 | ||
BC018910.2 | - | 4 | 929 | 3959973 | ||
BP314322.1 | - | 4 | 458 | |||
EU794638.1 | - | 4 | 904 |