Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194294.2 | NP_919270.2 | 3, 4 | 470 | ||
GenBank mRNA | AK128691.1 | - | 3, 4 | 470 | ||
BC113496.1 | - | 3, 4 | 249 | |||
BC113498.1 | - | 3, 4 | 249 | |||
EF052681.1 | - | 2, 3 | 189 |