Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135241.2 | NP_001128713.1 | 2 | 317 | ||
NM_001321027.1 | NP_001307956.1 | 2 | 317 | |||
NM_001354.5 | NP_001345.1 | 4 | 551 | |||
NM_205845.2 | NP_995317.1 | 3 | 451 | |||
GenBank mRNA | AB021654.1 | - | 2 | 138 | ||
AB031084.1 | - | 2 | 138 | |||
AB209017.1 | - | 2 | 205 | |||
AK091194.1 | - | 4 | 533 | |||
AK290304.1 | - | 2 | 194 | |||
AK296419.1 | - | 2 | 166 | |||
AK296686.1 | - | 2 | 164 | |||
AK308385.1 | - | 2 | 165 | |||
BC007024.1 | - | 2 | 160 | 3681970 | ||
BC063574.1 | - | 3 | 418 | 3877666 | ||
BT006653.1 | - | 2 | 138 | |||
DA438648.1 | - | 3 | 423 | |||
DA458030.1 | - | 4 | 446 | |||
M33376.1 | - | 2 | 93 | |||
S68330.1 | - | 2 | 144 | |||
U05598.1 | - | 2 | 161 |