Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002237.3 | NP_002228.2 | 2 | 612 | ||
XM_006723785.2 | XP_006723848.1 | 4 | 995 | |||
XM_011528800.1 | XP_011527102.1 | 3 | 670 | |||
XM_011528801.1 | XP_011527103.1 | 4 | 779 | |||
XM_011528802.1 | XP_011527104.1 | 4 | 774 | |||
XM_011528803.2 | XP_011527105.1 | 4 | 701 | |||
XM_011528804.1 | XP_011527106.1 | 3 | 924 | |||
XM_011528805.2 | XP_011527107.1 | 2 | 2835 | |||
XM_011528806.1 | XP_011527108.1 | 2 | 816 | |||
GenBank mRNA | AF033383.1 | - | 1 | 317 | ||
AK290689.1 | - | 2 | 613 | |||
BC006367.1 | - | 3 | 828 | 4131541 | ||
BC046629.2 | - | 2 | 581 | 5287759 | ||
CN301212.1 | - | 1 | 333 |