Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290224.1 | NP_001277153.1 | 19 | 2280 | ||
NM_199259.3 | NP_954868.1 | 23 | 2502 | |||
NM_199260.3 | NP_954869.1 | 22 | 2442 | |||
NM_199261.3 | NP_954870.2 | 24 | 2556 | |||
GenBank mRNA | AL833345.1 | - | 24 | 2463 |