Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001865.3 | NP_001856.2 | 4 | 469 | ||
NR_029466.1 | 4 | 522 | ||||
GenBank mRNA | BC100852.1 | - | 4 | 275 | 40002220 | |
BC100853.1 | - | 4 | 275 | 40002221 | ||
BC100854.2 | - | 4 | 275 | 40002222 | ||
BC133654.1 | - | 4 | 275 | 40002224 | ||
CB139775.1 | - | 4 | 381 | |||
CK821341.1 | - | 4 | 480 | |||
HE647861.1 | - | 4 | 469 | |||
X15822.1 | - | 4 | 288 |