Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016371.3 | NP_057455.1 | 6 | 783 | ||
GenBank mRNA | AF098786.2 | - | 6 | 695 | ||
AF145023.1 | - | 6 | 779 | |||
AJ249179.1 | - | 6 | 708 | |||
AK022929.1 | - | 6 | 1802 | |||
AK290741.1 | - | 6 | 752 | |||
AY358962.1 | - | 6 | 691 | |||
BC007068.1 | - | 6 | 739 | 3997494 | ||
BC065246.1 | - | 6 | 754 | 6062803 | ||
BT007075.1 | - | 6 | 684 |