Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001072.3 | NP_001063.2 | 1, 2 | 992 | ||
NM_205862.1 | NP_995584.1 | 2, 3 | 205 | |||
GenBank mRNA | AY435141.1 | - | 1, 2 | 853 | ||
BC019861.1 | - | 1, 2 | 887 | 4454322 | ||
BM688740.1 | - | 2, 3 | 205 | |||
BM924331.1 | - | 2, 3 | 223 | |||
DQ364250.1 | - | 1, 2 | 861 | |||
J04093.1 | - | 1, 2 | 937 |