Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199420.3 | NP_955452.3 | 4 | 717 | ||
XM_011512343.2 | XP_011510645.1 | 4 | 705 | |||
XM_011512347.2 | XP_011510649.1 | 4 | 705 | |||
XM_011512348.2 | XP_011510650.1 | 3 | 574 | |||
XM_011512352.2 | XP_011510654.1 | 4 | 705 | |||
GenBank mRNA | AB209867.1 | - | 4 | 645 | ||
AY032677.1 | - | 5 | 987 | |||
AY338826.2 | - | 4 | 717 | |||
BP365212.1 | - | 3 | 562 | |||
CR936627.1 | - | 3 | 563 |