Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010578.2 | NP_034708.1 | 4 | 501 | ||
GenBank mRNA | AK088016.1 | - | 4 | 402 | ||
AK088729.1 | - | 4 | 248 | |||
AK159689.1 | - | 4 | 343 | |||
AK167682.1 | - | 4 | 399 | |||
BC050906.1 | - | 4 | 374 | 5721348 | ||
X15202.1 | - | 3 | 194 | |||
Y00769.1 | - | 4 | 289 |