Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004815.3 | NP_004806.3 | 3 | 617 | ||
XM_006711048.3 | XP_006711111.1 | 2 | 251 | |||
XM_011542438.2 | XP_011540740.1 | 3 | 492 | |||
XM_011542439.2 | XP_011540741.1 | 3 | 337 | |||
XM_011542440.2 | XP_011540742.1 | 2 | 211 | |||
GenBank mRNA | AB208873.1 | - | 2 | 223 | ||
AK307733.1 | - | 3 | 446 | |||
AK310491.1 | - | 3 | 337 | |||
BC022483.1 | - | 3 | 499 | |||
BC067839.1 | - | 3 | 612 | |||
BC093741.1 | - | 2 | 290 | 7939586 | ||
BC093767.1 | - | 2 | 293 | 7939612 | ||
BI548487.1 | - | 3 | 620 | |||
U90920.1 | - | 3 | 431 |