Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004815.3 | NP_004806.3 | 9 | 1224 | ||
XM_006711048.3 | XP_006711111.1 | 8 | 858 | |||
XM_011542438.2 | XP_011540740.1 | 9 | 1099 | |||
XM_011542439.2 | XP_011540741.1 | 9 | 944 | |||
XM_011542440.2 | XP_011540742.1 | 8 | 818 | |||
GenBank mRNA | AB208873.1 | - | 8 | 830 | ||
AK307733.1 | - | 9 | 1053 | |||
AK310491.1 | - | 9 | 944 | |||
BC022483.1 | - | 9 | 1106 | |||
BC067839.1 | - | 9 | 1219 | |||
BC093741.1 | - | 8 | 897 | 7939586 | ||
BC093767.1 | - | 8 | 900 | 7939612 | ||
BX508890.1 | - | 2 | 154 | |||
U90920.1 | - | 9 | 1038 |