Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPCAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005768.5 | NP_005759.4 | 2 | 354 | ||
GenBank mRNA | AK096775.1 | - | 2 | 543 | ||
AK296164.1 | - | 2 | 290 | |||
AK296171.1 | - | 2 | 290 | |||
AU125262.1 | - | 2 | 426 | |||
BC000664.2 | - | 1 | 89 | |||
BC065194.1 | - | 2 | 285 | 5227816 | ||
BX647983.1 | - | 2 | 294 | |||
BX648009.1 | - | 2 | 325 | |||
U72515.1 | - | 1 | 39 |