Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNKSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297647.1 | NP_001284576.1 | 20 | 1791 | ||
NM_001297648.1 | NP_001284577.1 | 20 | 1765 | |||
NM_006314.2 | NP_006305.2 | 20 | 1770 | |||
XM_011540484.2 | XP_011538786.1 | 20 | 1750 | |||
XM_011540485.2 | XP_011538787.1 | 17 | 1280 | |||
GenBank mRNA | AB209335.1 | - | 20 | 1764 | ||
AF100153.1 | - | 20 | 1748 | |||
AK001900.1 | - | 20 | 1744 | |||
AK222817.1 | - | 20 | 1749 | |||
AK222904.1 | - | 20 | 1753 | |||
AK292646.1 | - | 20 | 1752 | |||
BC011604.2 | - | 20 | 1772 | 3139227 | ||
BC012797.2 | - | 20 | 1772 | 3505293 | ||
BT006900.1 | - | 20 | 1733 |