Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C1ORF61 SIRNA Assays ›
Gene Symbol:
C1orf61
Gene Name:
chromosome 1 open reading frame 61
Gene Aliases:
CROC4
Chromosome Location:
Chr.1: 156404252 - 156456649 on Build GRCh38
UniGene ID:
Hs.380027
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122599
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C1orf61
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320453.1
NP_001307382.1
5
529
NM_001320454.1
NP_001307383.1
4
531
NM_001320455.1
NP_001307384.1
4
482
NM_006365.2
NP_006356.1
7
781
NR_135260.1
4
433
NR_135261.1
5
518
NR_135262.1
3
333
NR_135263.1
5
567
NR_135264.1
6
685
NR_135265.1
2
297
NR_135266.1
3
370
NR_135267.1
3
397
NR_135268.1
7
1015
XM_011509067.1
XP_011507369.1
6
680
XM_011509068.1
XP_011507370.1
5
580
XM_011509070.1
XP_011507372.1
6
1094
XM_011509071.2
XP_011507373.1
6
1881
XM_011509072.1
XP_011507374.1
6
704
XM_011509073.2
XP_011507375.1
5
868
XM_011509074.1
XP_011507376.1
5
649
XM_017000062.1
XP_016855551.1
10
1262
XM_017000063.1
XP_016855552.1
8
886
XM_017000064.1
XP_016855553.1
6
803
XM_017000065.1
XP_016855554.1
8
953
XM_017000066.1
XP_016855555.1
8
1010
GenBank mRNA
AK091014.1
-
1
2316
AK094493.1
-
1
1781
AK124566.1
-
1
4513
AK124870.1
-
1
3337
BC016978.1
-
1
935
BC050448.1
-
5
432
BC050741.1
-
3
224
BC054045.1
-
4
464
BC059943.1
-
5
418
BC107686.1
-
4
292
BC128199.1
-
3
261
BC128200.1
-
3
261
BC141918.1
-
3
261
BC141919.1
-
3
261
BF347271.1
-
466
BG716287.1
-
5
409
BI597719.1
-
3
216
BI667681.1
-
4
508
BI668594.1
-
5
451
DB534652.2
-
4
460
DN990706.1
-
4
379
HY121703.1
-
2
298
HY137784.1
-
3
370
HY152089.1
-
3
397
U49857.1
-
7
625
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-ndlhf:80/100.66.76.213:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline