Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006369.4 | NP_006360.3 | 7 | 1981 | ||
XM_017000074.1 | XP_016855563.1 | 7 | 2007 | |||
GenBank mRNA | AF370430.1 | - | 7 | 1881 | ||
AK125827.1 | - | 6 | 3258 | |||
AK223420.1 | - | 4 | 1343 | |||
AK291283.1 | - | 7 | 1964 | |||
AK296436.1 | - | 7 | 1800 | |||
AK297596.1 | - | 7 | 1995 | |||
BC004953.1 | - | 4 | 1217 | |||
BC101522.1 | - | 7 | 1971 | 8069028 | ||
BC101524.1 | - | 7 | 1971 | 8069030 | ||
BX537366.1 | - | 7 | 1891 | |||
X86018.1 | - | 4 | 1193 |