Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006762.2 | NP_006753.1 | 8 | 1563 | ||
XM_011542098.2 | XP_011540400.1 | 6 | 1396 | |||
GenBank mRNA | AK292009.1 | - | 8 | 1513 | ||
D42042.1 | - | 8 | 1493 | |||
U51240.1 | - | 8 | 1547 |