Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007553.2 | NP_001007554.1 | 13 | 1886 | ||
NM_001130523.2 | NP_001123995.1 | 13 | 1940 | |||
NM_001242891.1 | NP_001229820.1 | 14 | 2033 | |||
NM_001242892.1 | NP_001229821.1 | 12 | 1499 | |||
NM_001242893.1 | NP_001229822.1 | 11 | 1406 | |||
NM_007158.5 | NP_009089.4 | 12 | 1793 | |||
GenBank mRNA | AB020692.1 | - | 13 | 1800 | ||
AF070542.1 | - | 11 | 1390 | |||
AF077054.1 | - | 12 | 1709 | |||
AK022516.1 | - | 11 | 1308 | |||
AK022528.1 | - | 11 | 1308 | |||
AK290146.1 | - | 12 | 1431 | |||
AL050150.1 | - | 2 | 86 | |||
AY049788.1 | - | 10 | 1295 | |||
BC032446.1 | - | 12 | 1728 | 5111213 | ||
BX648561.1 | - | 12 | 1630 | |||
BX648780.1 | - | 12 | 1727 | |||
CR749378.1 | - | 13 | 1890 | |||
CR749431.1 | - | 13 | 1872 |