Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272038.1 | NP_001258967.1 | 5, 6 | 666 | ||
NM_002870.3 | NP_002861.1 | 5, 6 | 548 | |||
NR_073553.1 | 5, 6 | 644 | ||||
XM_017001959.1 | XP_016857448.1 | 4, 5 | 301 | |||
GenBank mRNA | AF498948.1 | - | 5, 6 | 407 | ||
AK291580.1 | - | 5, 6 | 467 | |||
AY423722.1 | - | 5, 6 | 408 | |||
BC000799.1 | - | 5, 6 | 501 | 3451945 | ||
BC009227.2 | - | 1 | 384 | |||
BC073168.1 | - | 5, 6 | 452 | 6193572 | ||
BC094846.1 | - | 5, 6 | 532 | |||
BT019700.1 | - | 5, 6 | 407 | |||
BT019701.1 | - | 5, 6 | 407 | |||
X75593.1 | - | 5, 6 | 546 |