Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323623.1 | NP_001310552.1 | 6, 7 | 798 | ||
NM_001323624.1 | NP_001310553.1 | 5, 6 | 613 | |||
NM_013285.2 | NP_037417.1 | 6, 7 | 798 | |||
GenBank mRNA | AK298358.1 | - | 6, 7 | 708 | ||
AK312915.1 | - | 6, 7 | 752 | |||
BC000107.2 | - | 6, 7 | 734 | 3506131 | ||
BC009250.2 | - | 6, 7 | 734 | 2960125 | ||
L05425.1 | - | 6, 7 | 714 |