Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NENF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013349.4 | NP_037481.1 | 2, 3 | 292 | ||
GenBank mRNA | AB126219.1 | - | 2, 3 | 235 | ||
AF049672.1 | - | 2, 3 | 90 | |||
AF173937.1 | - | 2, 3 | 295 | |||
AK074431.1 | - | 2, 3 | 260 | |||
AK223135.1 | - | 2, 3 | 260 | |||
AY762102.1 | - | 2, 3 | 251 | |||
BC008823.2 | - | 2, 3 | 241 | 4303614 |