Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NENF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013349.4 | NP_037481.1 | 4 | 904 | ||
NR_026598.1 | 3 | 843 | ||||
GenBank mRNA | BC008823.2 | - | 4 | 853 | 4303614 | |
DB574171.1 | - | 1 | 414 |