Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286067.1 | NP_001272996.1 | 10 | 1298 | ||
NM_001286068.1 | NP_001272997.1 | 9 | 1161 | |||
NM_001286069.1 | NP_001272998.1 | 9 | 1166 | |||
NM_001286070.1 | NP_001272999.1 | 8 | 1014 | |||
NM_001286071.1 | NP_001273000.1 | 8 | 989 | |||
NM_014039.3 | NP_054758.2 | 8 | 1016 | |||
XM_006718824.1 | XP_006718887.1 | 10 | 1207 | |||
XM_011542781.2 | XP_011541083.1 | 9 | 1480 | |||
XM_011542782.2 | XP_011541084.1 | 9 | 1571 | |||
XM_017017613.1 | XP_016873102.1 | 10 | 4622 | |||
XM_017017614.1 | XP_016873103.1 | 10 | 1449 | |||
XM_017017615.1 | XP_016873104.1 | 10 | 1902 | |||
XM_017017616.1 | XP_016873105.1 | 8 | 1008 | |||
XM_017017617.1 | XP_016873106.1 | 8 | 1404 | |||
XM_017017618.1 | XP_016873107.1 | 8 | 980 | |||
XM_017017619.1 | XP_016873108.1 | 7 | 862 | |||
GenBank mRNA | AK129932.1 | - | 4 | 585 | ||
AK225977.1 | - | 9 | 1096 | |||
AK291833.1 | - | 9 | 1072 | |||
AK292215.1 | - | 10 | 1217 | |||
AL136605.1 | - | 9 | 1092 | |||
AY203960.1 | - | 8 | 1042 | |||
BC007110.1 | - | 8 | 921 | |||
BC012298.1 | - | 8 | 983 | 3852558 | ||
CR533538.1 | - | 8 | 934 |