Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286067.1 | NP_001272996.1 | 10 | 2445 | ||
NM_001286068.1 | NP_001272997.1 | 9 | 2308 | |||
NM_001286069.1 | NP_001272998.1 | 9 | 2313 | |||
NM_001286070.1 | NP_001272999.1 | 8 | 2161 | |||
NM_001286071.1 | NP_001273000.1 | 8 | 2136 | |||
NM_014039.3 | NP_054758.2 | 8 | 2163 | |||
XM_006718824.1 | XP_006718887.1 | 10 | 2354 | |||
XM_011542781.2 | XP_011541083.1 | 9 | 2627 | |||
XM_011542782.2 | XP_011541084.1 | 9 | 2718 | |||
XM_017017613.1 | XP_016873102.1 | 10 | 5769 | |||
XM_017017614.1 | XP_016873103.1 | 10 | 2596 | |||
XM_017017615.1 | XP_016873104.1 | 10 | 3049 | |||
XM_017017616.1 | XP_016873105.1 | 8 | 2155 | |||
XM_017017619.1 | XP_016873108.1 | 7 | 2009 | |||
GenBank mRNA | AK129932.1 | - | 4 | 1729 | ||
AY203960.1 | - | 8 | 2188 | |||
BC012298.1 | - | 8 | 2130 | 3852558 |