Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MACROD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014067.3 | NP_054786.2 | 3 | 547 | ||
XM_005273939.4 | XP_005273996.1 | 3 | 1086 | |||
XM_005273940.2 | XP_005273997.1 | 3 | 1086 | |||
XM_006718521.3 | XP_006718584.1 | 3 | 1086 | |||
XM_006718522.2 | XP_006718585.1 | 3 | 1086 | |||
XM_011544970.2 | XP_011543272.1 | 3 | 1086 | |||
GenBank mRNA | AF086206.1 | - | 3 | 167 | ||
AF202922.2 | - | 3 | 480 | |||
BC000270.1 | - | 3 | 495 | 3357931 | ||
BC003188.1 | - | 3 | 301 | 3502054 | ||
BC007297.1 | - | 3 | 495 | 3349763 | ||
BC008316.1 | - | 3 | 495 | 3342973 |