Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIEXF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014388.6 | NP_055203.4 | 5, 6 | 750 | ||
XM_006711275.3 | XP_006711338.1 | 5, 6 | 750 | |||
XM_011509418.2 | XP_011507720.1 | 5, 6 | 774 | |||
GenBank mRNA | AK289613.1 | - | 5, 6 | 730 | ||
AK314061.1 | - | 5, 6 | 780 | |||
BC022964.1 | - | 5, 6 | 711 | 5110673 | ||
BX345520.2 | - | 5, 6 | 736 | |||
BX648514.1 | - | 5, 6 | 663 | |||
CR749825.1 | - | 5, 6 | 687 |