Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIEXF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014388.6 | NP_055203.4 | 10 | 1863 | ||
XM_011509418.2 | XP_011507720.1 | 10 | 1887 | |||
GenBank mRNA | AK096118.1 | - | 3 | 943 | ||
AK123674.1 | - | 4 | 669 | |||
AK289613.1 | - | 10 | 1843 | |||
AK314061.1 | - | 10 | 1893 | |||
BC022964.1 | - | 10 | 1824 | 5110673 | ||
BX648514.1 | - | 10 | 1776 | |||
CR749825.1 | - | 10 | 1800 |