Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACYBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007214.1 | NP_001007215.1 | 2 | 454 | ||
NM_014412.2 | NP_055227.1 | 2 | 639 | |||
XM_017001046.1 | XP_016856535.1 | 2 | 586 | |||
GenBank mRNA | AF057356.1 | - | 2 | 328 | ||
AF275803.1 | - | 2 | 273 | |||
AF314752.1 | - | 2 | 338 | |||
AK093425.1 | - | 2 | 426 | |||
AK294042.1 | - | 2 | 353 | |||
AK313278.1 | - | 2 | 430 | |||
AL035305.1 | - | 2 | 331 | |||
AY423723.1 | - | 2 | 215 | |||
BC005975.1 | - | 2 | 440 | 4102572 | ||
BC022352.1 | - | 2 | 337 | 4281339 | ||
BC067823.1 | - | 2 | 321 | 5260416 | ||
BC078151.1 | - | 2 | 304 | 6065872 | ||
BQ432793.1 | - | 2 | 457 |