Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014458.3 | NP_055273.2 | 3 | 747 | ||
XM_005245093.3 | XP_005245150.1 | 4 | 896 | |||
XM_011509420.2 | XP_011507722.1 | 3 | 776 | |||
XM_017001052.1 | XP_016856541.1 | 3 | 1026 | |||
XM_017001053.1 | XP_016856542.1 | 3 | 1026 | |||
XM_017001054.1 | XP_016856543.1 | 4 | 896 | |||
GenBank mRNA | AB026190.1 | - | 3 | 688 | ||
AJ844466.1 | - | 2 | 568 | |||
AK001430.1 | - | 3 | 704 | |||
AK308048.1 | - | 2 | 665 | |||
BC005253.1 | - | 3 | 698 | |||
BC063418.1 | - | 3 | 677 | 5221643 |