Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142685.1 | NP_001136157.1 | 19 | 2103 | ||
NM_014715.3 | NP_055530.2 | 10 | 1435 | |||
XM_005271736.4 | XP_005271793.2 | 20 | 3133 | |||
XM_011543072.2 | XP_011541374.1 | 20 | 2335 | |||
XM_011543073.2 | XP_011541375.2 | 20 | 2071 | |||
XM_011543075.2 | XP_011541377.1 | 11 | 1281 | |||
XM_017018595.1 | XP_016874084.1 | 20 | 2052 | |||
XM_017018596.1 | XP_016874085.1 | 18 | 2329 | |||
GenBank mRNA | AB018255.2 | - | 10 | 1356 | ||
AB088416.1 | - | 10 | 1356 | |||
AL833062.2 | - | 18 | 1905 | |||
AY194287.1 | - | 10 | 1359 | |||
BC104898.1 | - | 9 | 1090 | 8143901 | ||
BC113429.1 | - | 9 | 1090 | 8322481 | ||
EF127492.1 | - | 19 | 2103 |