Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATG13 SIRNA Assays ›
Gene Symbol:
ATG13
Gene Name:
autophagy related 13
Gene Aliases:
KIAA0652, PARATARG8
Chromosome Location:
Chr.11: 46617276 - 46676019 on Build GRCh38
UniGene ID:
Hs.127403
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122700
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142673.2
NP_001136145.1
14
1733
NM_001205119.1
NP_001192048.1
14
1776
NM_001205120.1
NP_001192049.1
13
1677
NM_001205121.1
NP_001192050.1
13
1617
NM_001205122.1
NP_001192051.1
11
1484
NM_014741.4
NP_055556.2
13
1622
XM_005253262.2
XP_005253319.1
15
1572
XM_005253263.2
XP_005253320.1
15
1863
XM_005253265.2
XP_005253322.1
16
2069
XM_005253266.2
XP_005253323.1
15
1798
XM_005253268.2
XP_005253325.1
13
1175
XM_006718394.2
XP_006718457.1
16
1919
XM_006718395.2
XP_006718458.1
16
1854
XM_006718396.2
XP_006718459.1
15
1953
XM_011520493.1
XP_011518795.1
17
2056
XM_011520496.2
XP_011518798.1
14
1831
XM_011520499.2
XP_011518801.1
14
1445
XM_011520500.1
XP_011518802.1
16
1892
XM_011520501.1
XP_011518803.1
15
1820
XM_011520502.1
XP_011518804.1
15
1793
XM_011520503.1
XP_011518805.1
14
1709
XM_017018599.1
XP_016874088.1
16
2004
XM_017018600.1
XP_016874089.1
15
1545
XM_017018601.1
XP_016874090.1
14
1720
XM_017018602.1
XP_016874091.1
15
1970
XM_017018603.1
XP_016874092.1
16
2087
XM_017018604.1
XP_016874093.1
14
1764
XM_017018605.1
XP_016874094.1
15
1755
XM_017018606.1
XP_016874095.1
14
1699
XM_017018607.1
XP_016874096.1
12
1076
XM_017018608.1
XP_016874097.1
14
1446
XM_017018609.1
XP_016874098.1
14
1737
XM_017018610.1
XP_016874099.1
13
1621
XM_017018611.1
XP_016874100.1
14
1672
XM_017018612.1
XP_016874101.1
14
1859
XM_017018613.1
XP_016874102.1
13
1653
XM_017018614.1
XP_016874103.1
14
1644
XM_017018615.1
XP_016874104.1
13
1743
XM_017018616.1
XP_016874105.1
12
1537
XM_017018617.1
XP_016874106.1
14
1742
XM_017018618.1
XP_016874107.1
16
2058
XM_017018619.1
XP_016874108.1
14
1843
XM_017018620.1
XP_016874109.1
14
1903
XM_017018621.1
XP_016874110.1
13
1787
XM_017018622.1
XP_016874111.1
13
1744
GenBank mRNA
AB014552.1
-
13
1357
AK023867.1
-
13
1355
AK223603.1
-
14
1452
AK289641.1
-
14
1280
AK293109.1
-
13
1321
AK294110.1
-
11
1197
AK296526.1
-
6
564
AK315925.1
-
6
491
BC001331.1
-
13
1051
3458359
BC002378.2
-
13
1321
2961127
BC006191.2
-
13
1363
3936851
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
macroautophagy
mitophagy in response to mitochondrial depolarization
positive regulation of protein targeting to mitochondrion
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline