Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014810.4 | NP_055625.4 | 3 | 474 | ||
XM_011510191.2 | XP_011508493.1 | 3 | 433 | |||
XM_011510198.2 | XP_011508500.1 | 3 | 433 | |||
XM_017002929.1 | XP_016858418.1 | 3 | 433 | |||
XM_017002930.1 | XP_016858419.1 | 3 | 471 | |||
XM_017002931.1 | XP_016858420.1 | 3 | 433 | |||
GenBank mRNA | AF287356.1 | - | 3 | 258 | ||
AF387614.1 | - | 2 | 88 | |||
AK095601.1 | - | 3 | 459 | |||
DA716735.1 | - | 3 | 472 |