Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014847.3 | NP_055662.3 | 27 | 3876 | ||
XM_005245658.3 | XP_005245715.1 | 28 | 3960 | |||
XM_005245667.3 | XP_005245724.1 | 27 | 3909 | |||
XM_011510215.2 | XP_011508517.1 | 28 | 3957 | |||
XM_011510216.1 | XP_011508518.1 | 28 | 3852 | |||
XM_017002972.1 | XP_016858461.1 | 28 | 3982 | |||
XM_017002973.1 | XP_016858462.1 | 28 | 3849 | |||
XM_017002974.1 | XP_016858463.1 | 28 | 3880 | |||
XM_017002976.1 | XP_016858465.1 | 27 | 3811 | |||
XM_017002978.1 | XP_016858467.1 | 27 | 3832 | |||
XM_017002979.1 | XP_016858468.1 | 27 | 3793 | |||
GenBank mRNA | AJ243669.2 | - | 10 | 1482 | ||
BC003170.1 | - | 26 | 3758 | 2906083 |