Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242925.1 | NP_001229854.1 | 4 | 1733 | ||
NM_001297611.1 | NP_001284540.1 | 3 | 1191 | |||
NM_001297613.1 | NP_001284542.1 | 3 | 1211 | |||
NM_014858.3 | NP_055673.2 | 4 | 2397 | |||
XM_005245684.1 | XP_005245741.1 | 4 | 2088 | |||
XM_005245685.4 | XP_005245742.1 | 4 | 1719 | |||
XM_005245686.3 | XP_005245743.1 | 4 | 2114 | |||
XM_017003000.1 | XP_016858489.1 | 3 | 1172 | |||
GenBank mRNA | AB001596.1 | - | 3 | 1067 | ||
AB007950.3 | - | 4 | 2397 | |||
AK094913.1 | - | 1 | 257 | |||
AK095816.1 | - | 3 | 1211 | |||
AK131419.1 | - | 5 | 2186 | |||
AK293734.1 | - | 4 | 1733 | |||
AK314398.1 | - | 3 | 1090 | |||
AY358192.1 | - | 3 | 2305 | |||
BC041019.1 | - | 3 | 1131 | |||
BC053876.1 | - | 855 | ||||
BC131629.1 | - | 4 | 1869 | 40118729 |