Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXPH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144763.1 | NP_001138235.1 | 6 | 907 | ||
NM_001144764.1 | NP_001138236.1 | 4 | 642 | |||
NM_001144765.1 | NP_001138237.1 | 3 | 593 | |||
NM_001308019.1 | NP_001294948.1 | 7 | 824 | |||
NM_015065.2 | NP_055880.2 | 6 | 951 | |||
XM_011542696.2 | XP_011540998.1 | 7 | 845 | |||
XM_011542698.2 | XP_011541000.1 | 6 | 1291 | |||
XM_011542700.2 | XP_011541002.1 | 6 | 2191 | |||
XM_017017397.1 | XP_016872886.1 | 7 | 840 | |||
XM_017017398.1 | XP_016872887.1 | 7 | 837 | |||
XM_017017399.1 | XP_016872888.1 | 6 | 743 | |||
XM_017017400.1 | XP_016872889.1 | 7 | 2259 | |||
XM_017017401.1 | XP_016872890.1 | 6 | 2188 | |||
XM_017017402.1 | XP_016872891.1 | 7 | 2299 | |||
XM_017017403.1 | XP_016872892.1 | 4 | 1445 | |||
XM_017017404.1 | XP_016872893.1 | 5 | 2142 | |||
GenBank mRNA | AB014524.1 | - | 7 | 824 | ||
AK303993.1 | - | 4 | 622 | |||
AK304212.1 | - | 3 | 594 | |||
AY099469.1 | - | 6 | 937 | |||
BC113119.1 | - | 6 | 922 | 40037748 | ||
BC117701.1 | - | 6 | 922 | 40074153 | ||
CR627226.1 | - | 6 | 913 |