Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290184.1 | NP_001277113.1 | 17 | 2193 | ||
NM_001290186.1 | NP_001277115.1 | 13 | 1640 | |||
NM_015331.2 | NP_056146.1 | 16 | 2054 | |||
GenBank mRNA | AF240468.1 | - | 16 | 2072 | ||
AK296153.1 | - | 11 | 1339 | |||
AK299142.1 | - | 13 | 1527 | |||
AK314764.1 | - | 16 | 1961 | |||
AY359120.1 | - | 16 | 1931 | |||
BC047621.1 | - | 17 | 2150 | 4827966 | ||
D87442.1 | - | 16 | 1928 |